4-Pentenoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Allylacetic acid, Pent-4-enoic acid

002777-25G 002777-100G
FLUO002777-25GEA 0 GBP
FLUO002777-25G FLUO002777-100G
4-Pentenoic acid
4-Pentenoic acid
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 187
Melting Pt: –18 °C
Density: 0.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004408
CAS Number: 591-80-0
EINECS: 209-732-7
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR