Methyl oleate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: cis-Oleic Acid Methyl Ester, Oleic acid methyl ester

M-Clarity™ quality level = MQ200

311111-5G 311111-25G
SIAL311111-5GEA 134 GBP
SIAL311111-5G SIAL311111-25G
Methyl oleate, Sigma-Aldrich®
Methyl oleate
Formula: C₁₉H₃₆O₂
MW: 296.49 g/mol
Boiling Pt: ∼218.5 °C (1013 hPa)
Melting Pt: –20 °C
Density: 0.88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009578
CAS Number: 112-62-9
EINECS: 203-992-5
Merck Index: 12,06965

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR