1,1,2-Trichloro-3,3,3-trifluoropropene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1,1,2-Trichlorotrifluoropropene, 1,1,2-Trichloro-3,3,3-trifluoro-1-propene
007453-5G
007453-25G
007453-100G
FLUO007453-5GEA
0
GBP
FLUO007453-5G
FLUO007453-25G
FLUO007453-100G
1,1,2-Trichloro-3,3,3-trifluoropropene
1,1,2-Trichloro-3,3,3-trifluoropropene
Formula:
C₃Cl₃F₃ Boiling Pt: 88 °C (1013 hPa) Density: 1.618 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013672 CAS Number: 431-52-7 EINECS: 207-075-0 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...