Trichloroethylene
Supplier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
1,1,2-Trichloroethene, Trichloroethene, 1,1,2-Trichloroethylene
9579.1
9579.2
9579.3
ROTH9579.1EA
0
GBP
ROTH9579.1
ROTH9579.2
ROTH9579.3
Trichloroethylene
Trichloroethylene
Formula:
ClCH=CCl₂ MW: 131.39 g/mol Boiling Pt: 86.7 Melting Pt: –73 °C Density: 1.46 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000838 CAS Number: 79-01-6 EINECS: 201-167-4 UN: 1710 ADR: 6.1 III Merck Index: 12,09769 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...