trans-2-Octenal, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: (E)-2-Octenal

M-Clarity™ quality level = MQ100

269956-25G 269956-5G
SIAL269956-25GEA 235 GBP
SIAL269956-25G SIAL269956-5G
trans-2-Octenal, Sigma-Aldrich®
trans-2-Octenal
Formula: C₈H₁₄O
MW: 126.2 g/mol
MDL Number: MFCD00007011
CAS Number: 2548-87-0
EINECS: 219-833-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR