trans-2-Dodecenal, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: (E)-2-Dodecenal

M-Clarity™ quality level = MQ300

W240206-100G-K W240206-1KG-K W240206-5KG-K
SIALW240206-100G-KEA 103 GBP
SIALW240206-100G-K SIALW240206-1KG-K SIALW240206-5KG-K
trans-2-Dodecenal, Sigma-Aldrich®
trans-2-Dodecenal
Formula: C₁₂H₂₂O
MW: 182.31 g/mol
MDL Number: MFCD00014674
CAS Number: 20407-84-5
EINECS: 243-797-2
UN: 1760
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR