(3-Fluorophenyl)acetic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001293-25G 001293-100G 001293-250G
FLUO001293-25GEA 0 GBP
FLUO001293-25G FLUO001293-100G FLUO001293-250G
(3-Fluorophenyl)acetic acid
(3-Fluorophenyl)acetic acid
Formula: (C₆H₄F)CH₂COOH
Boiling Pt: 100…120 °C (0.5 mmHg)
Melting Pt: 44…45.2 °C
MDL Number: MFCD00004331
CAS Number: 331-25-9
EINECS: 206-360-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR