(3,4-Difluorophenyl)acetic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3,4-Difluorophenylacetic acid

002092-5G 002092-25G 002092-100G
FLUO002092-5GEA 0 GBP
FLUO002092-5G FLUO002092-25G FLUO002092-100G
(3,4-Difluorophenyl)acetic acid
(3,4-Difluorophenyl)acetic acid
Formula: (C₆H₃F₂)CH₂COOH
MW: 172.13 g/mol
Boiling Pt: 259
Melting Pt: 40…42 °C
MDL Number: MFCD00010002
CAS Number: 658-93-5
EINECS: 211-527-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR