(2,6-Difluorophenyl)acetic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2,6-Difluorophenylacetic acid

001966-5G 001966-25G
FLUO001966-5GEA 0 GBP
FLUO001966-5G FLUO001966-25G
(2,6-Difluorophenyl)acetic acid
(2,6-Difluorophenyl)acetic acid
Formula: (C₆H₃F₂)CH₂COOH
MW: 172.13 g/mol
Boiling Pt: 259
Melting Pt: 100…102 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010001
CAS Number: 85068-28-6
EINECS: 285-289-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR