(2,5-Difluorophenyl)acetic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2,5-Difluorophenylacetic acid

001944.1G 001944-5G 001944-25G
FLUO001944.1GEA 0 GBP
FLUO001944.1G FLUO001944-5G FLUO001944-25G
(2,5-Difluorophenyl)acetic acid
(2,5-Difluorophenyl)acetic acid
Formula: (C₆H₃F₂)CH₂COOH
MW: 172.13 g/mol
Boiling Pt: 259
Melting Pt: 122…128 °C
MDL Number: MFCD00010000
CAS Number: 85068-27-5
EINECS: 285-288-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR