(2,3,4-Trifluorophenyl)acetic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-(2,3,4-Trifluorophenyl)acetic acid

007690-1G 007690-5G
FLUO007690-1GEA 0 GBP
FLUO007690-1G FLUO007690-5G
(2,3,4-Trifluorophenyl)acetic acid
(2,3,4-Trifluorophenyl)acetic acid
Formula: C₆H₂F₃CH₂COOH
MW: 190.12 g/mol
MDL Number: MFCD00061235
CAS Number: 243666-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR