(2,3-Dimethoxyphenyl)acetic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(2,3-Dimethoxyphenyl)acetic acid

018593-1G 018593-5G
FLUO018593-1GEA 0 GBP
FLUO018593-1G FLUO018593-5G
(2,3-Dimethoxyphenyl)acetic acid
(2,3-Dimethoxyphenyl)acetic acid
Formula: (C₆H₃(OCH₃)₂)CH₂COOH
MW: 196.2 g/mol
MDL Number: MFCD00060351
CAS Number: 90-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR