(4-Hydroxy-3-nitrophenyl)acetic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-(4-Hydroxy-3-nitrophenyl)acetic acid

018724-1G 018724-25G 018724-5G
FLUO018724-1GEA 0 GBP
FLUO018724-1G FLUO018724-25G FLUO018724-5G
(4-Hydroxy-3-nitrophenyl)acetic acid
(4-Hydroxy-3-nitrophenyl)acetic acid
Formula: (C₆H₃(OH)(NO₂))CH₂COOH
MW: 197.15 g/mol
Melting Pt: 149…150 °C
Storage Temperature: Freezer
MDL Number: MFCD00007122
CAS Number: 10463-20-4
EINECS: 233-948-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR