Methyl phenylacetate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Phenylacetic acid methyll ester

M-Clarity™ quality level = MQ300

W273309-10KG-K W273309-1KG-K W273309-5KG-K
SIALW273309-10KG-KEA 470 GBP
SIALW273309-10KG-K SIALW273309-1KG-K SIALW273309-5KG-K
Methyl phenylacetate, Sigma-Aldrich®
Methyl phenylacetate
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 218 °C (1013 hPa)
Melting Pt: 49…50 °C
Density: 1.067 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008453
CAS Number: 101-41-7
EINECS: 202-940-9
Merck Index: 12,07422

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR