(2-Chloro-6-fluorophenyl)acetic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

004460-5G 004460-25G 004460-10G 004460-1G
FLUO004460-5GEA 0 GBP
FLUO004460-5G FLUO004460-25G FLUO004460-10G FLUO004460-1G
(2-Chloro-6-fluorophenyl)acetic acid
(2-Chloro-6-fluorophenyl)acetic acid
Formula: (C₆H₃ClF)CH₂COOH
Boiling Pt: 289 °C (1013 hPa)
Melting Pt: 120…123 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004319
CAS Number: 37777-76-7
EINECS: 253-661-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR