4-Nitrophenyl chloroformate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ200

160210-100G 160210-25G 160210-5G
SIAL160210-100GEA 252 GBP
SIAL160210-100G SIAL160210-25G SIAL160210-5G
4-Nitrophenyl chloroformate, Sigma-Aldrich®
4-Nitrophenyl chloroformate
Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
Boiling Pt: 159 °C (19 mmHg)
Melting Pt: 78…81 °C
MDL Number: MFCD00007321
CAS Number: 7693-46-1
EINECS: 231-706-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR