(3,4-Dichlorophenyl)acetic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3,4-Dichlorophenylacetic acid
M-Clarity™ quality level = MQ100
280003-25G
280003-5G
SIAL280003-25GEA
111
GBP
SIAL280003-25G
SIAL280003-5G
(3,4-Dichlorophenyl)acetic acid, Sigma-Aldrich®
(3,4-Dichlorophenyl)acetic acid
Formula:
(C₆H₃Cl₂)CH₂COOH MW: 205.04 g/mol Boiling Pt: 314 °C (1013 hPa) Melting Pt: 91…92 °C |
MDL Number:
MFCD00004333 CAS Number: 5807-30-7 EINECS: 227-368-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...