(2,4-Dichlorophenyl)acetic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: DCAA, 2,4-Dichlorophenylacetic acid

001382-25G 001382-100G
FLUO001382-25GEA 0 GBP
FLUO001382-25G FLUO001382-100G
(2,4-Dichlorophenyl)acetic acid
(2,4-Dichlorophenyl)acetic acid
Formula: (C₆H₃Cl₂)CH₂COOH
Boiling Pt: 316 °C (1013 hPa)
Melting Pt: 132…133 °C
MDL Number: MFCD00004318
CAS Number: 19719-28-9
EINECS: 243-248-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR