(2-Bromophenyl)acetic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-(2-Bromophenyl)acetic acid
001133-5G
001133-25G
FLUO001133-5GEA
0
GBP
FLUO001133-5G
FLUO001133-25G
(2-Bromophenyl)acetic acid
(2-Bromophenyl)acetic acid
Formula:
(C₆H₄Br)CH₂COOH MW: 215.05 g/mol Boiling Pt: 308 °C (1013 hPa) Melting Pt: 107.5 °C |
MDL Number:
MFCD00004314 CAS Number: 18698-97-0 EINECS: 242-509-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...