(α,α,α-Trifluoro-4-tolyl)acetic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
002185-5G
002185-25G
FLUO002185-5GEA
0
GBP
FLUO002185-5G
FLUO002185-25G
(α,α,α-Trifluoro-4-tolyl)acetic acid
(α,α,α-Trifluoro-4-tolyl)acetic acid
Formula:
(C₆H₄(CF₃))CH₂COOH |
MDL Number:
MFCD00004352 CAS Number: 32857-62-8 EINECS: 251-263-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...