(3,5-Bis(trifluoromethyl)phenyl)acetic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
002105-5G
002105-100G
002105-25G
FLUO002105-5GEA
0
GBP
FLUO002105-5G
FLUO002105-100G
FLUO002105-25G
(3,5-Bis(trifluoromethyl)phenyl)acetic acid
(3,5-Bis(trifluoromethyl)phenyl)acetic acid
Formula:
(C₆H₃(CF₃)₂)CH₂COOH Boiling Pt: 276 °C (1013 hPa) Melting Pt: 121…125 °C |
MDL Number:
MFCD00009908 CAS Number: 85068-33-3 EINECS: 285-295-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...