(α,α,α-Trifluoro-2-tolyl)acetic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001978-1G 001978-5G 001978-25G
FLUO001978-1GEA 0 GBP
FLUO001978-1G FLUO001978-5G FLUO001978-25G
(α,α,α-Trifluoro-2-tolyl)acetic acid
(α,α,α-Trifluoro-2-tolyl)acetic acid
Formula: F₃CC₆H₄CH₂COOH
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 98…100 °C
MDL Number: MFCD00004327
CAS Number: 3038-48-0
EINECS: 221-240-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR