(2,4-Bis(trifluoromethyl)phenyl)acetic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2,4-Bis(trifluoromethyl)phenylacetic acid

007830-1G 007830-5G
FLUO007830-1GEA 0 GBP
FLUO007830-1G FLUO007830-5G
(2,4-Bis(trifluoromethyl)phenyl)acetic acid
(2,4-Bis(trifluoromethyl)phenyl)acetic acid
Formula: (C₆H₃(CF₃)₂)OCH₂COOH
MW: 272.15 g/mol
MDL Number: MFCD00042492
CAS Number: 177952-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR