Isopropyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

008986-10G 008986-50G
FLUO008986-10GEA 0 GBP
FLUO008986-10G FLUO008986-50G
Isopropyl isothiocyanate
Isopropyl isothiocyanate
Formula: C₄H₇NS
Boiling Pt: 136…137 °C (1013 hPa)
Density: 0.9475 g/cm³ (20 °C)
MDL Number: MFCD00037830
CAS Number: 2253-73-8
EINECS: 218-851-3
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR