4-Methylvaleric acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Isocaproic acid, 4-Methylpentanoic acid

002770-5G 002770-25G
FLUO002770-5GEA 0 GBP
FLUO002770-5G FLUO002770-25G
4-Methylvaleric acid
4-Methylvaleric acid
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: –35 °C
Density: 0,9192 g/cm³ (20 °C)
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00002803
CAS Number: 646-07-1
EINECS: 211-464-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR