Isobutyric acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Methylpropionic acid, 2-Methylpropanoic acid

M-Clarity™ quality level = MQ300

W222208-10KG-K W222208-1KG-K W222208-25KG-K W222208-5KG-K
SIALW222208-10KG-KEA 308 GBP
SIALW222208-10KG-K SIALW222208-1KG-K SIALW222208-25KG-K SIALW222208-5KG-K
Isobutyric acid, Sigma-Aldrich®
Isobutyric acid
Formula: C₄H₈O₂
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: –45 °C
Density: 0.949 g/cm³ (20 °C)
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00002658
CAS Number: 79-31-2
EINECS: 201-195-7
UN: 2529
ADR: 3,III
Merck Index: 13,05172

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR