(±)-2-Methylcaproic acid ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
2-MethylhexanoicAcid, 2-Methylhexanoic acid, (±)-2-Methylhexanoic acid
A11290.06
A11290.14
A11290.06EA
22.5
GBP
A11290.06
A11290.14
(±)-2-Methylcaproic acid ≥98%
(±)-2-Methylcaproic acid
Purity: 98%
CAS No.: 4536-23-6
Molecular Formula: C7H14O2
Formula Weight: 130.19
Flash Point: 105°(221°F)
Refractive Index: 1.4205
Odour: Rancid
Boiling Point: 209-210°
Beilstein Reference: 1721227
UN No.: 3265
MDL No.: MFCD00002674
Formula:
C₇H₁₄O₂ MW: 130.19 g/mol Boiling Pt: 209…210 °C Density: 0.916 Flash Pt: 105 °C (221 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002674 CAS Number: 4536-23-6 EINECS: 224-883-9 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...