2-Ethylcaproic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Ethylhexanoic acid

MQ200

E29141-1L E29141-2.5L E29141-2L E29141-500ML
SIALE29141-1LEA 117 GBP
SIALE29141-1L SIALE29141-2.5L SIALE29141-2L SIALE29141-500ML
2-Ethylcaproic acid, Sigma-Aldrich®
2-Ethylcaproic acid
Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 228
Melting Pt: –82.8 °C
Density: 0.91 g/cm³ (20 °C)
Flash Pt: 114
Storage Temperature: Ambient
MDL Number: MFCD00002675
CAS Number: 149-57-5
EINECS: 205-743-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR