2-Ethylbutyric acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Ethylbutanoic acid, Diethylacetic acid

M-Clarity™ quality level = MQ300

W242918-1KG W242918-25KG W242918-5KG W242918-SAMPLE
SIALW242918-1KGEA 285 GBP
SIALW242918-1KG SIALW242918-25KG SIALW242918-5KG SIALW242918-SAMPLE
2-Ethylbutyric acid, Sigma-Aldrich®
2-Ethylbutyric acid
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 193…194 °C (1013 hPa)
Melting Pt: –14 °C
Density: 0.92 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002670
CAS Number: 88-09-5
EINECS: 201-796-4
UN: 2810
ADR: 6.1,III
Merck Index: 13,03137

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR