2-Ethylbutyric acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Ethylbutanoic acid, Diethylacetic acid
M-Clarity™ quality level = MQ100
109959-100ML
109959-1L
SIAL109959-100MLEA
44.3
GBP
SIAL109959-100ML
SIAL109959-1L
2-Ethylbutyric acid, Sigma-Aldrich®
2-Ethylbutyric acid
Formula:
C₆H₁₂O₂ MW: 116.16 g/mol Boiling Pt: 193…194 °C (1013 hPa) Melting Pt: –14 °C Density: 0.92 g/cm³ (20 °C) Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002670 CAS Number: 88-09-5 EINECS: 201-796-4 UN: 2810 ADR: 6.1,III Merck Index: 13,03137 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...