L(-)-Glutathione (oxidised form), Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
L(-)-Glutathione, oxidized, Bi(glutathion-S-yl), GSSG, Glutathiol, Oxidized Glutathione (GSSG), Glutathione oxidized, (2S,2'S)-5,5'-(((2R,2'R)-Disulphanediylbis(1-((carboxymethyl)amino)-1-oxopropane-3,2-diyl))bis(azanediyl))bis(2-amino-5-oxopentanoic acid)
M-Clarity™ quality level = MQ200
G6654-1G
G6654-250MG
G6654-500MG
G6654-5G
SIALG6654-1GEA
360
GBP
SIALG6654-1G
SIALG6654-250MG
SIALG6654-500MG
SIALG6654-5G
L(-)-Glutathione (oxidised form), Sigma-Aldrich®
L(-)-Glutathione (oxidized form)
Formula:
C₂₀H₃₂N₆O₁₂S₂ MW: 612.64 g/mol Melting Pt: 185 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00063106 CAS Number: 27025-41-8 EINECS: 248-170-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...