2,6-Dimethyl-4-heptanone, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Diisobutyl Ketone
M-Clarity™ quality level = MQ100
273848-100G
273848-25G
SIAL273848-100GEA
482
GBP
SIAL273848-100G
SIAL273848-25G
2,6-Dimethyl-4-heptanone, Sigma-Aldrich®
2,6-Dimethyl-4-heptanone
Formula:
C₉H₁₈O MW: 142.24 g/mol Boiling Pt: 165 °C (1013 hPa) Melting Pt: –46 °C Density: 0.806 g/cm³ (20 °C) Flash Pt: 60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008940 CAS Number: 108-83-8 EINECS: 203-620-1 UN: 1224 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...