n-Triacontane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

263842-1G 263842-5G
SIAL263842-1GEA 93.8 GBP
SIAL263842-1G SIAL263842-5G
n-Triacontane, Sigma-Aldrich®
n-Triacontane
Formula: H₃C(CH₂)₂₈CH₃
MW: 422.82 g/mol
Boiling Pt: 449.7
Melting Pt: 65…67 °C
Density: 0.7797 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009410
CAS Number: 638-68-6
EINECS: 211-349-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR