n-Nonadecane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N-nonadecane

M-Clarity™ quality level = MQ100

N28906-100G N28906-25G
SIALN28906-100GEA 168 GBP
SIALN28906-100G SIALN28906-25G
n-Nonadecane, Sigma-Aldrich®
n-Nonadecane
Formula: H₃C(CH₂)₁₇CH₃
MW: 268.53 g/mol
Boiling Pt: 329.7 °C (1013 hPa)
Melting Pt: 32.5 °C
Density: 0.7845 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009012
CAS Number: 629-92-5
EINECS: 211-116-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR