n-Hexacosane, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
N-hexacosane
MQ100
241687-5G
241687-1G
SIAL241687-5GEA
104
GBP
SIAL241687-5G
SIAL241687-1G
n-Hexacosane, Sigma-Aldrich®
n-Hexacosane
Formula:
H₃C(CH₂)₂₄CH₃ MW: 366.71 g/mol Boiling Pt: 412.2 Melting Pt: 56…59 °C Density: 0.804 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009354 CAS Number: 630-01-3 EINECS: 211-124-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...