1-Octanesulphonic acid sodium salt, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Sodium octane-1-sulphonate

M-Clarity™ quality level = MQ100

O8380-100G O8380-25G O8380-5G O-8380 O8380-25G
SIALO8380-100GEA 668 GBP
SIALO8380-100G SIALO8380-25G SIALO8380-5G PFIZO-8380 NOVSO8380-25G
1-Octanesulphonic acid sodium salt, Sigma-Aldrich®
1-Octanesulphonic acid sodium salt
Formula: C₈H₁₇NaO₃S
MW: 216.28 g/mol
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007544
CAS Number: 5324-84-5
EINECS: 226-195-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1244cb11db07775dec2e193823e40080024b7036&vid=32d95cd9e321818e2f94c8261c4b4e6180ee0fc7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR