1-Hexanesulphonic acid sodium salt
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Sodium hexane-1-sulphonate, Sodium hexanesulphonate, Sodium 1-hexanesulphonate
009636-5G
009636-25G
FLUO009636-5GEA
0
GBP
FLUO009636-5G
FLUO009636-25G
1-Hexanesulphonic acid sodium salt
1-Hexanesulphonic acid sodium salt
Formula:
C₆H₁₃O₃SNa Melting Pt: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007542 CAS Number: 2832-45-3 EINECS: 220-601-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...