1-Dodecansulphonic acid sodium salt

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009635-5G 009635-25G
FLUO009635-5GEA 0 GBP
FLUO009635-5G FLUO009635-25G
1-Dodecansulphonic acid sodium salt
1-Dodecansulphonic acid sodium salt
Formula: C₁₂H₂₅NaO₃S
Melting Pt: 300 °C
Density: 1.218 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007527
CAS Number: 2386-53-0
EINECS: 219-200-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR