Methyl methanesulphonate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
129925-5G
129925-100G
129925-25G
SIAL129925-5GEA
66.2
GBP
SIAL129925-5G
SIAL129925-100G
SIAL129925-25G
Methyl methanesulphonate, Sigma-Aldrich®
Methyl methanesulphonate
Formula:
C₂H₆O₃S MW: 110.13 g/mol Boiling Pt: 203 Melting Pt: 20 Density: 1.3 g/cm³ (20 °C) Flash Pt: 104 Storage Temperature: Ambient |
MDL Number:
MFCD00007557 CAS Number: 66-27-3 EINECS: 200-625-0 UN: 2810 ADR: 6.1 III Merck Index: 12,06170 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...