Ethyl methanesulphonate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001758-25G 001758-100G
FLUO001758-25GEA 0 GBP
FLUO001758-25G FLUO001758-100G
Ethyl methanesulphonate
Ethyl methanesulphonate
Formula: C₃H₈O₃S
Boiling Pt: 213 °C (1013 hPa)
Density: 1.2079 g/cm³ (20 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00007559
CAS Number: 62-50-0
EINECS: 200-536-7
UN: 2810
ADR: 6.1,II
Merck Index: 13,03859

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR