Dimethyl sulphate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

41610-100ML 41610-500ML
SIAL41610-100MLEA 51.6 GBP
SIAL41610-100ML SIAL41610-500ML
Dimethyl sulphate, Sigma-Aldrich®
Dimethyl sulphate
Formula: C₂H₆O₄S
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: –31.8 °C
Density: 1.33 g/cm³ (20 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00008416
CAS Number: 77-78-1
EINECS: 201-058-1
UN: 1595
ADR: 6.1,I
Merck Index: 12,03305

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR