2-Mercaptoethanol

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2-Hydroxyethyl mercaptan, β-Mercaptoethanol

39563.01 39563.02
SERA39563.01EA 20.93 GBP
SERA39563.01 SERA39563.02
2-Mercaptoethanol
2-Mercaptoethanol
Formula: HSCH₂CH₂OH
MW: 78.14 g/mol
Boiling Pt: 157
Melting Pt: –100 °C
Density: 1.12 g/cm³ (20 °C)
Flash Pt: 68
Storage Temperature: Ambient
MDL Number: MFCD00004890
CAS Number: 60-24-2
EINECS: 200-464-6
UN: 2966
ADR: 6.1 II
Merck Index: 13,05894
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc427209b84d0343a5ae47c435ddfc53df4013d2&vid=7d70fd087d948d54e7a946d3b2310e2d820d2396&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR