Potassium methanolate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Potassium methoxide, Potassium methylate

M-Clarity™ quality level = MQ200

292788-250G 292788-5G
SIAL292788-250GEA 216 GBP
SIAL292788-250G SIAL292788-5G
Potassium methanolate, Sigma-Aldrich®
Potassium methanolate
Formula: CH₃KO
MW: 70.13 g/mol
Density: 0.85 g/cm³ (20 °C)
MDL Number: MFCD00012416
CAS Number: 865-33-8
EINECS: 212-736-1
UN: 3206
ADR: 4.2,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR