1-Tridecanol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1-Tridecanol

MQ200

T57630-10G T57630-50G
SIALT57630-10GEA 113 GBP
SIALT57630-10G SIALT57630-50G
1-Tridecanol, Sigma-Aldrich®
1-Tridecanol
Formula: H₃C(CH₂)₁₁CH₂OH
MW: 200.36 g/mol
Boiling Pt: 250
Melting Pt: 31.5
Density: 0.8215 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004756
CAS Number: 112-70-9
EINECS: 203-998-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR