1-Tetracosanol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

L3507-100MG L3507-1G L3507-250MG L3507-25MG
SIALL3507-100MGEA 275 GBP
SIALL3507-100MG SIALL3507-1G SIALL3507-250MG SIALL3507-25MG
1-Tetracosanol, Sigma-Aldrich®
1-Tetracosanol
Formula: H₃C(CH₂)₂₂CH₂OH
Boiling Pt: 190 °C (1013 hPa)
Melting Pt: 81…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00042662
CAS Number: 506-51-4
EINECS: 208-043-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR