1-Heptanol, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ300
W254800-1KG-K
W254800-20KG-K
W254800-4KG-K
SIALW254800-1KG-KEA
78
GBP
SIALW254800-1KG-K
SIALW254800-20KG-K
SIALW254800-4KG-K
1-Heptanol, Sigma-Aldrich®
1-Heptanol
Formula:
H₃C(CH₂)₅CH₂OH Boiling Pt: 176 °C (1013 hPa) Melting Pt: –35 °C Density: 0.82 g/cm³ (20 °C) Flash Pt: 70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002986 CAS Number: 111-70-6 EINECS: 203-897-9 Merck Index: 13,04681 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...