1-Heptanol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Heptyl alcohol

M-Clarity™ quality level = MQ200

H2805-1L H2805-2.5L H2805-250ML
SIALH2805-1LEA 113 GBP
SIALH2805-1L SIALH2805-2.5L SIALH2805-250ML
1-Heptanol, Sigma-Aldrich®
1-Heptanol
Formula: H₃C(CH₂)₅CH₂OH
MW: 116.2 g/mol
Boiling Pt: 176
Melting Pt: –35 °C
Density: 0.82 g/cm³ (20 °C)
Flash Pt: 70
Storage Temperature: Ambient
MDL Number: MFCD00002986
CAS Number: 111-70-6
EINECS: 203-897-9
Merck Index: 13,04681

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR