2-Nitropropane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ200

130265-5ML 130265-100ML 130265-2.5L 130265-500ML
SIAL130265-5MLEA 47.5 GBP
SIAL130265-5ML SIAL130265-100ML SIAL130265-2.5L SIAL130265-500ML
2-Nitropropane, Sigma-Aldrich®
2-Nitropropane
Formula: C₃H₇NO₂
Boiling Pt: 120.3 °C (1013 hPa)
Melting Pt: –91 °C
Density: 0.9927 g/cm³ (20 °C)
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00007397
CAS Number: 79-46-9
EINECS: 201-209-1
UN: 2608
ADR: 3,III
Merck Index: 12,06725

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR