1-Nitropropane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ100

N22851-100ML N22851-500ML
SIALN22851-100MLEA 33.3 GBP
SIALN22851-100ML SIALN22851-500ML
1-Nitropropane, Sigma-Aldrich®
1-Nitropropane
Formula: C₃H₇NO₂
Boiling Pt: 131 °C (1013 hPa)
Melting Pt: –108 °C
Density: 0.9934 g/cm³ (20 °C)
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00007407
CAS Number: 108-03-2
EINECS: 203-544-9
UN: 2608
ADR: 3,III
Merck Index: 12,06724

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR