2-Ethylaminoethanol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N-Ethyl-2-aminoethanol, N-Ethyl-2-ethanolamin, 2-(Ethylamino)ethan-1-ol

M-Clarity™ quality level = MQ200

471461-100ML 471461-2L 471461-500ML
SIAL471461-100MLEA 59 GBP
SIAL471461-100ML SIAL471461-2L SIAL471461-500ML
2-Ethylaminoethanol, Sigma-Aldrich®
2-Ethylaminoethanol
Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 167
Melting Pt: –9 °C
Density: 0.914 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002841
CAS Number: 110-73-6
EINECS: 203-797-5
UN: 2922
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR